Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Ethoxyethoxy) Ethyl Acetate, 99%, Spectrum™ Chemical
SDP

Catalog No. 18608140 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-140 20 L Amber Glass Bottle
18-608-138 1 L Amber Glass Bottle
18-608-139 4 L Amber Glass Bottle
18-608-141 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-608-140 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. E102320LTMT
Add to Cart
Add to Cart

Chemical Identifiers

CAS 112-15-2
Molecular Formula C8H16O4
Molecular Weight (g/mol) 176.21
InChI Key FPZWZCWUIYYYBU-UHFFFAOYSA-N
IUPAC Name 2-(2-ethoxyethoxy)ethyl acetate
SMILES CCOCCOCCOC(C)=O

Specifications

Acidity 0.0002
CAS Max % 1
Boiling Point 215°C (419°F)
Packaging Amber Glass Bottle
Quantity 20 L
Specific Gravity 1.008 - 1.013
Percent Purity 0.99
Grade Reagent
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy) Ethyl Acetate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.