Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Ethoxyethoxy)ethyl Acrylate (stabilized with MEHQ) 98.0+%, TCI America™
SDP

Catalog No. p-7129087
Click to view available options
Quantity:
25 g
500 g

Chemical Identifiers

CAS 7328-17-8
Molecular Formula C9H16O4
Molecular Weight (g/mol) 188.223
InChI Key FTALTLPZDVFJSS-UHFFFAOYSA-N
Synonym Acrylic Acid 2-(2-Ethoxyethoxy)ethyl Ester, Diethylene Glycol Monoethyl Ether Acrylate
PubChem CID 81766
IUPAC Name 2-(2-ethoxyethoxy)ethyl prop-2-enoate
SMILES CCOCCOCCOC(=O)C=C

Specifications

Color Colorless
Boiling Point 250°C
Quantity 25 g
Formula Weight 188.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2-(2-Ethoxyethoxy)ethyl Acrylate (stabilized with MEHQ)
EINECSNumber (2)-3106
RTECSNumber UD3343500
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.