Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(2-[2-(Fmoc-amino)ethoxy]ethoxy)acetic acid, 95%

Catalog No. p-7055137
Click to view available options
Quantity:
250 mg
1 g

CAS: 166108-71-0 | C21H23NO6 | 385.416 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 166108-71-0
Molecular Formula C21H23NO6
Molecular Weight (g/mol) 385.416
MDL Number MFCD01321015
InChI Key XQPYRJIMPDBGRW-UHFFFAOYSA-N
Synonym 2-2-fmoc-amino ethoxy ethoxy acetic acid, fmoc-nh-peg2-ch2cooh, fmoc-aeea, 1-9h-fluoren-9-yl-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, 8-fmoc-amino-3,6-dioxaoctanoic acid, fmoc-adoa, fmoc-aeeac-oh, fmoc-amino-3,6 dioxaoctanoic acid, 8-fmoc-amino-3,6-dioxa-n-octanoic acid
PubChem CID 2756092
IUPAC Name 2-[2-[2-(9H-fluoren-9-ylmethoxycarbonylamino)ethoxy]ethoxy]acetic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NCCOCCOCC(=O)O

Specifications

Color White
Linear Formula FmocNHCH2CH2OCH2CH2OCH2CO2H
Quantity 250 mg
Beilstein 9658732
Formula Weight 385.42
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (2-[2-(Fmoc-amino)ethoxy]ethoxy)acetic acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.