Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Furoin, 98%

Catalog No. AAA1527214
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 552-86-3 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 552-86-3
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00003246
InChI Key MIJRFWVFNKQQDK-UHFFFAOYNA-N
Synonym furoin, 2,2'-furoin, 1,2-di furan-2-yl-2-hydroxyethanone, furoylfurylcarbinol, ethanone, 1,2-di-2-furanyl-2-hydroxy, alpha-furoin, .alpha.-furoin, ethanone, 1,2-di-2-furyl-2-hydroxy, 1,2-bis furan-2-yl-2-hydroxyethan-1-one, 1,2-di-2-furanyl-2-hydroxyethanone
PubChem CID 11100
IUPAC Name 1,2-bis(furan-2-yl)-2-hydroxyethanone
SMILES OC(C(=O)C1=CC=CO1)C1=CC=CO1

Specifications

Melting Point 137°C to 139°C
Quantity 25 g
Beilstein 383995
Formula Weight 192.17
Percent Purity 98%
Chemical Name or Material 2,2'-Furoin
EINECSNumber 209-024-8
RTECSNumber KM5774095
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.