Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Hydroxyphenyl)benzothiazole 98.0+%, TCI America™
SDP

Catalog No. H097325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H097325G 25 g
H09735G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H097325G Supplier TCI America Supplier No. H097325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3411-95-8
Molecular Formula C13H9NOS
Molecular Weight (g/mol) 227.28
MDL Number MFCD00022869
InChI Key NSDGEQWRYXOZLN-UKTHLTGXSA-N
Synonym phenol, o-2-benzothiazolyl, 2-2-hydroxyphenyl benzothiazole, o-2-benzothiazolyl phenol, phenol, 2-2-benzothiazolyl, 2-2-benzothiazolyl phenol, 2-o-hydroxyphenyl benzothiazole, 2-2'-hydroxyphenyl benzothiazole, 2-1,3-benzothiazol-2-yl phenol, 6z-6-3h-1,3-benzothiazol-2-ylidene cyclohexa-2,4-dien-1-one, mvvgspcxhrfddr-uhfffaoysa-n
PubChem CID 5376551
IUPAC Name 6-[(2E)-2,3-dihydro-1,3-benzothiazol-2-ylidene]cyclohexa-2,4-dien-1-one
SMILES O=C1C=CC=C\C1=C1\NC2=CC=CC=C2S1

Specifications

Melting Point 133°C
Quantity 25 g
Formula Weight 227.28
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Hydroxyphenyl)benzothiazole
RTECSNumber SJ7360000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.