Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methyl-1,3-thiazol-4-yl)ethanol, 97%, Thermo Scientific™

Catalog No. p-4812638
Click to view available options
Quantity:
250 mg
1 g

Chemical Identifiers

CAS 121357-04-8
Molecular Formula C6H9NOS
Molecular Weight (g/mol) 143.204
InChI Key REKVZAFNJAMAQL-UHFFFAOYSA-N
Synonym 2-2-methyl-1,3-thiazol-4-yl ethanol, 2-2-methylthiazol-4-yl ethanol, 2-2-methyl-1,3-thiazol-4-yl ethan-1-ol, 4-thiazoleethanol,2-methyl, 2-2-methyl-4-thiazolyl ethanol, acmc-20duqt, 2-methyl-4-thiazoleethanol, 2-methylthiazole-4-ethanol, 4-hydroxyethyl-2-methyl-1,3-thiazole
PubChem CID 19876739
IUPAC Name 2-(2-methyl-1,3-thiazol-4-yl)ethanol
SMILES CC1=NC(=CS1)CCO

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 143.21
Chemical Name or Material 2-(2-Methyl-1, 3-thiazol-4-yl)ethanol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.