Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine 98.0+%, TCI America™
SDP

Catalog No. M25841G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M25841G 1 g
M25845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M25841G Supplier TCI America Supplier No. M25841G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 152460-09-8
Molecular Formula C16H13N5O2
Molecular Weight (g/mol) 307.313
MDL Number MFCD02179269
InChI Key OJITWRFPRCHSMX-UHFFFAOYSA-N
Synonym n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimidin-2-amine, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidine-amine, chembl20426, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidineamine, n-2-methyl-5-nitrophenyl-4-pyridin-3-ylpyrimidin-2-amine, 2-methyl-4-nitro-phenyl-4-pyridin-3-yl-pyrimidin-2-yl-amine, 2-methyl-5-nitro-n-4-pyridin-3-yl pyrimidin-2-yl benzenamine, 2-pyrimidinamine, n-2-methyl-5-nitrophenyl-4-3-pyridinyl, n-2-methyl-5-nitrophenyl-4-pyridin-3-yl pyrimid, n-2-methyl-5-nitrophenyl-4-3-pyridyl-2-pyrimidinamine
PubChem CID 10686149
IUPAC Name N-(2-methyl-5-nitrophenyl)-4-pyridin-3-ylpyrimidin-2-amine
SMILES CC1=C(C=C(C=C1)[N+](=O)[O-])NC2=NC=CC(=N2)C3=CN=CC=C3

Specifications

Melting Point 194°C
Color Yellow
Quantity 1 g
Formula Weight 307.31
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2-(2-Methyl-5-nitroanilino)-4-(3-pyridyl)pyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.