Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(2-Propynyloxy)ethylamine 98.0+%, TCI America™
SDP

Catalog No. P21661G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P21661G 1 g
P21665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P21661G Supplier TCI America Supplier No. P21661G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 122116-12-5
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
InChI Key MZFRRBQTCNHISM-UHFFFAOYSA-N
PubChem CID 18356755
IUPAC Name 2-prop-2-ynoxyethanamine
SMILES C#CCOCCN

Specifications

Color Yellow
Quantity 1 g
UN Number 2734
Formula Weight 99.13
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 2-(2-Propynyloxy)ethylamine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.