Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sulfuryl chloride, 98.5%

Catalog No. AC169450025
Click to view available options
Quantity:
1 kg
2.5 kg
Packaging:
Glass bottle

CAS: 7791-25-5 | Cl2O2S | 134.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7791-25-5
Molecular Formula Cl2O2S
Molecular Weight (g/mol) 134.96
MDL Number MFCD00011451
InChI Key YBBRCQOCSYXUOC-UHFFFAOYSA-N
Synonym sulfuryl chloride, sulfonyl chloride, sulphuryl dichloride, sulphuryl chloride, sulfonyl dichloride, sulfuric dichloride, sulfuric oxychloride, sulfurylchlorid, caswell no. 816, sulfurylchloride
PubChem CID 24648
ChEBI CHEBI:29291
IUPAC Name sulfuryl dichloride
SMILES ClS(Cl)(=O)=O

Specifications

Boiling Point 69.1°C
Melting Point -54.1°C
Color Colorless to Yellow
Physical Form Liquid
Quantity 2.5 kg
Assay Percent Range 98.5%
Linear Formula SO2Cl2
Fieser 01,1128; 02,394; 03,276; 04,474; 05,641; 06,561; 10,375; 13,284; 14,291; 16,311
Solubility Information Solubility in water: reacts. Other solubilities: soluble in glacial acetic acid
Formula Weight 134.96
Percent Purity 98.5%
Packaging Glass bottle
Refractive Index 1.443
Specific Gravity 1.667
Density 1.6670g/mL
Chemical Name or Material Sulfuryl chloride, 98.5%
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Reacts violently with water.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-245-6
RTECSNumber WT4870000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.