Alfa Aesar™ 2,2'-Thiodiethanethiol, 90+%

Manufacturer: Alfa Aesar™ B2230430

Quantity 250g

 View more versions of this product

Catalog No. AAB2230430

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3570-55-6
    Molecular Formula HSCH2CH2SCH2CH2SH
    Formula Weight 154.32
    MDL Number MFCD00004893
    Synonym Bis(2-mercaptoethyl) sulfide; 2-Mercaptoethyl sulfide
    Chemical Name or Material 2,2'-Thiodiethanethiol

    Specifications

    Formula Weight 154.32
    MDL Number MFCD00004893
    Synonym Bis(2-mercaptoethyl) sulfide; 2-Mercaptoethyl sulfide
    Health Hazard 2 GHS H Statement
    H332-H315-H319-H335-H227
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.
    Flash Point 90°C (194°F)
    Boiling Point 135° to 136°C (10mmHg)
    Quantity 250g
    TSCA TSCA
    Molecular Formula HSCH2CH2SCH2CH2SH
    CAS 3570-55-6
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Chemical Name or Material 2,2'-Thiodiethanethiol
    Assay ≥90%

    Description

    Applications
    2,2 ft.-Thiodiethanol is used in the synthesis of novel luminescent polynuclear gold(I) phosphine complexes 2,2 ft.-Thiodiethanethiol is used for extraction photometric determination of Ni, gives color reaction with metals.

    Solubility
    Difficult to mix.

    Notes
    Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

    GHS H Statement
    H332-H315-H319-H335-H227
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Combustible liquid.

    GHS P Statement
    P210-P261-P280-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit