Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,2'-Bipyridine, 1% (w/v) in Methanol, Ricca Chemical
SDP

Catalog No. R09900001C Shop All Ricca Chemical Company Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
R09900001C 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. R09900001C Supplier Ricca Chemical Company Supplier No. R09900001C
Add to Cart
Add to Cart

Chemical Identifiers

CAS 67-56-1 , 366-18-7
Molecular Formula CH4O
Molecular Weight (g/mol) Mixture
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
IUPAC Name methanol
SMILES CO

Specifications

Boiling Point Approximately 64.5°C
Odor Odorless
Assay Percent Range 1% (w/v) in Methanol
Packaging Amber Glass Bottle
Quantity 1 L
Solubility Information Solubility in Water: Infinite
Percent Purity 1% w/v in Methanol
Grade Laboratory
Physical Form Liquid
Recommended Storage Flammable
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.