Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,5,6-Pentafluorobenzyl alcohol, 96-98%, Thermo Scientific™

Catalog No. AC188360050
Click to view available options
:
5g

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 440-60-8
CAS Min % 96.0
CAS Max % 100.0
Infrared Spectrum Authentic
Assay Percent Range 96-98%
Packaging Glass bottle
Molecular Formula C7H3F5O
Linear Formula C6F5CH2OH
Quantity 5g
MDL Number MFCD00004602
Synonym 2,3,4,5,6-pentafluorobenzyl alcohol, pentafluorobenzyl alcohol, perfluorophenyl methanol, pentafluorophenyl methanol, 2,3,4,5,6-pentafluorophenyl methanol, 2,3,4,5,6-pentafluorobenzenemethanol, hydroxymethyl pentafluorobenzene, benzenemethanol, 2,3,4,5,6-pentafluoro, pentafluorobenzylalcohol, 2,3,4,5,6-pentafluorobenzylalcohol
InChI Key PGJYYCIOYBZTPU-UHFFFAOYSA-N
SMILES C(C1=C(C(=C(C(=C1F)F)F)F)F)O
IUPAC Name (2,3,4,5,6-pentafluorophenyl)methanol
Molecular Weight (g/mol) 198.092
PubChem CID 9923
ChEBI CHEBI:44903
Formula Weight 198.09
Percent Purity ≥96%
Chemical Name or Material 2,3,4,5,6-Pentafluorobenzyl alcohol
Show More Show Less
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.