Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetrafluorobenzyl Alcohol 98.0+%, TCI America™
SDP

Catalog No. T19425G
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 53001-70-0
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD03701105
InChI Key BXLQTYVTBJQPPZ-UHFFFAOYSA-N
PubChem CID 17750859
IUPAC Name (2,3,4,6-tetrafluorophenyl)methanol
SMILES OCC1=C(F)C=C(F)C(F)=C1F

Specifications

Color Yellow
Quantity 5 g
Formula Weight 180.10
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,4,6-Tetrafluorobenzyl Alcohol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.