Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4,6-Tetrafluoropyridine, 98+%, Thermo Scientific™

Catalog No. AAL1954514
Click to view available options
Quantity:
25 g

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 3512-13-8
Boiling Point 88°C to 90°C
Molecular Formula C5HF4N
Refractive Index 1.399
Quantity 25 g
MDL Number MFCD03001159
UN Number UN1760
Beilstein 1368002
Synonym pyridine, 2,3,4,6-tetrafluoro, acmc-20aoin, pubchem22302, 2,3,4,6-tetrakis fluoranyl pyridine
InChI Key NOQFPTRRIPJRMG-UHFFFAOYSA-N
SMILES C1=C(C(=C(N=C1F)F)F)F
IUPAC Name 2,3,4,6-tetrafluoropyridine
Molecular Weight (g/mol) 151.064
PubChem CID 2783279
Formula Weight 151.06
Percent Purity ≥98%
Chemical Name or Material 2,3,4,6-Tetrafluoropyridine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.