Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,4-Trimethoxybenzaldehyde, 98+%

Catalog No. AAA1060918
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 2103-57-3 | C10H12O4 | 196.202 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2103-57-3
Molecular Formula C10H12O4
Molecular Weight (g/mol) 196.202
MDL Number MFCD00003310
InChI Key UCTUXUGXIFRVGX-UHFFFAOYSA-N
Synonym trimethoxybenzaldehyde, benzaldehyde, 2,3,4-trimethoxy, 2,3,4-trimethoxy-benzaldehyde, benzaldehyde, trimethoxy, 2,3,4 trimethoxybenzylaldehyde, pubchem8261, trimethoxy benzaldehyde, trimetazidine impurity c, acmc-1b07v, ksc201q5r
PubChem CID 75006
IUPAC Name 2,3,4-trimethoxybenzaldehyde
SMILES COC1=C(C(=C(C=C1)C=O)OC)OC

Specifications

Melting Point 38°C to 42°C
Boiling Point 168°C to 170°C (12 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.5547
Quantity 50 g
Beilstein 981091
Sensitivity Air Sensitive
Formula Weight 196.2
Percent Purity ≥98%
Chemical Name or Material 2,3,4-Trimethoxybenzaldehyde
Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 218-271-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.