Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluoro-p-toluic acid, 98%, Thermo Scientific™

Catalog No. AC356100050
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 652-32-4
Molecular Formula C8H4F4O2
Molecular Weight (g/mol) 208.11
MDL Number MFCD00012302
InChI Key COOULIOYEXBFDT-UHFFFAOYSA-N
Synonym 2,3,5,6-tetrafluoro-p-toluic acid, 4-methyl-2,3,5,6-tetrafluorobenzoic acid, p-toluic acid, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluoro-4-methyl-benzoic acid, tetrafluoro-p-toluic acid, acmc-1az53, 2,3,5,6-tetrafluoro-4-methylbenzoicacid, 2,3,5,6-tetrafluoro-4-toluic acid, 4-carboxy-2,3,5,6-tetrafluorotoluene
PubChem CID 69548
IUPAC Name 2,3,5,6-tetrafluoro-4-methylbenzoic acid
SMILES CC1=C(C(=C(C(=C1F)F)C(=O)O)F)F

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 168°C to 172°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 208.11
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 2,3,5,6-Tetrafluoro-p-toluic acid, 98%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.