Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3,5,6-Tetrafluorobenzyl alcohol, 98%

Click to view available options
Quantity:
1g
5g
25g
4084-38-2
C7H4F4O
180.102
MFCD00792428
AGWVQASYTKCTCC-UHFFFAOYSA-N
2,3,5,6-tetrafluorobenzyl alcohol,2,3,5,6-tetrafluorophenyl methanol,2,3,5,6-tetrafluorobenzylalcohol,benzenemethanol, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorophenyl methan-1-ol,2,3,5,6-tetrafluorobenyl alcohol,pubchem15529,acmc-209jh7,2,3,5,6-tetrafluorbenzyl alcohol,benzenemethanol,2,3,5,6-tetrafluoro
2734029
(2,3,5,6-tetrafluorophenyl)methanol
C1=C(C(=C(C(=C1F)F)CO)F)F
4084-38-2
C7H4F4O
180.102
MFCD00792428
AGWVQASYTKCTCC-UHFFFAOYSA-N
2,3,5,6-tetrafluorobenzyl alcohol,2,3,5,6-tetrafluorophenyl methanol,2,3,5,6-tetrafluorobenzylalcohol,benzenemethanol, 2,3,5,6-tetrafluoro,2,3,5,6-tetrafluorophenyl methan-1-ol,2,3,5,6-tetrafluorobenyl alcohol,pubchem15529,acmc-209jh7,2,3,5,6-tetrafluorbenzyl alcohol,benzenemethanol,2,3,5,6-tetrafluoro
2734029
(2,3,5,6-tetrafluorophenyl)methanol
C1=C(C(=C(C(=C1F)F)CO)F)F
Odor Recognizable
Melting Point 34°C to 38°C
Quantity 5g
Boiling Point 220°C to 225°C
Formula Weight 180.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl alcohol

GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.

P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c

H302-H317-H319

EINECSNumber : 443-840-7

TSCA : No

Recommended Storage : Ambient temperatures