Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorobenzyl alcohol, 98%

Catalog No. AAB2516214
Click to view available options
Quantity:
5 g
25 g

CAS: 4084-38-2 | C7H4F4O | 180.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4084-38-2
Molecular Formula C7H4F4O
Molecular Weight (g/mol) 180.10
MDL Number MFCD00792428
InChI Key AGWVQASYTKCTCC-UHFFFAOYSA-N
PubChem CID 2734029
IUPAC Name (2,3,5,6-tetrafluorophenyl)methanol
SMILES OCC1=C(F)C(F)=CC(F)=C1F

Specifications

Melting Point 34°C to 38°C
Boiling Point 220°C to 225°C
Odor Recognizable
Quantity 25 g
Synonym 2,3,5,6-tetrafluorobenzyl alcohol, 2,3,5,6-tetrafluorophenyl methanol, 2,3,5,6-tetrafluorobenzylalcohol, benzenemethanol, 2,3,5,6-tetrafluoro, 2,3,5,6-tetrafluorophenyl methan-1-ol, 2,3,5,6-tetrafluorobenyl alcohol, pubchem15529, acmc-209jh7, 2,3,5,6-tetrafluorbenzyl alcohol, benzenemethanol,2,3,5,6-tetrafluoro
Formula Weight 180.1
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorobenzyl alcohol
Hazard Category H302-H317-H319
Hazard Statement GHS H Statement
H301-H319-H317
Toxic if swallowed.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P363-P501c
EINECSNumber 443-840-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.