Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5,6-Tetrafluorophenol, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
769-39-1
C6H2F4O
166.07
MFCD00002157
PBYIIRLNRCVTMQ-UHFFFAOYSA-N
phenol, 2,3,5,6-tetrafluoro,unii-tna118i5tg,2,3,5,6-tetrafluoro phenol,tna118i5tg,2,3,5,6-tetrafluor-phenol,ccris 5843,zlchem 411,pubchem2135,acmc-1bc5w,2,3,5,6tetrafluorophenol
69858
2,3,5,6-tetrafluorophenol
C1=C(C(=C(C(=C1F)F)O)F)F
769-39-1
C6H2F4O
166.07
MFCD00002157
PBYIIRLNRCVTMQ-UHFFFAOYSA-N
phenol, 2,3,5,6-tetrafluoro,unii-tna118i5tg,2,3,5,6-tetrafluoro phenol,tna118i5tg,2,3,5,6-tetrafluor-phenol,ccris 5843,zlchem 411,pubchem2135,acmc-1bc5w,2,3,5,6tetrafluorophenol
69858
2,3,5,6-tetrafluorophenol
C1=C(C(=C(C(=C1F)F)O)F)F
Boiling Point 140°C
Flash Point 79°C
Melting Point 33°C to 38°C
Packaging Glass bottle
Color White
Infrared Spectrum Authentic
Quantity 5g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula HC6F4OH
Formula Weight 166.07
Physical Form Crystalline Low Melting Solid
Percent Purity 98%
Chemical Name or Material 2,3,5,6-Tetrafluorophenol, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 212-209-6

TSCA : TSCA