Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Collidine, 99%

Catalog No. AAB2353714
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 695-98-7 | C8H11N | 121.183 g/mol

2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,3,5-Collidine is omeprazole intermediate which is the drug involved in proton pump inhibitors to block the production of stomach acid. It is used in the manufacture vitamin D3.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 695-98-7
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.183
MDL Number MFCD00126840
InChI Key GFYHSKONPJXCDE-UHFFFAOYSA-N
Synonym 2,3,5-collidine, pyridine, 2,3,5-trimethyl, 2,3,5-trimethyl pyridine, pubchem15901, acmc-1aul1, 2,3,5-trimethyl-pyridin, 2,3,5-trimethyl-pyridine, ksc490s0f
PubChem CID 12759
IUPAC Name 2,3,5-trimethylpyridine
SMILES CC1=CC(=C(N=C1)C)C

Specifications

Density 0.935
Boiling Point 182°C to 184°C
Flash Point 74°C (165°F)
Odor Unpleasant
Refractive Index 1.5075
Quantity 25 g
UN Number UN2810
Beilstein 108832
Solubility Information Slightly soluble in water.
Formula Weight 121.18
Percent Purity 99%
Chemical Name or Material 2,3,5-Collidine
Show More Show Less
Hazard Category H226-H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H227-H302-H312-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Hazard Class: 3; Packaging Group: III
EINECSNumber 211-786-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.