Alfa Aesar™ 2,3,5-Trimethylphenol, 98+%

Manufacturer: Alfa Aesar™ A1074522

Quantity 100g

 View more versions of this product

Catalog No. AAA1074522

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 697-82-5
    Molecular Formula C9H12O
    Formula Weight 136.2
    MDL Number MFCD00002228
    UN Number UN2430
    Beilstein 2042210
    Synonym Isopseudocumenol
    Chemical Name or Material 2,3,5-Trimethylphenol

    Specifications

    Formula Weight 136.2
    Health Hazard 3 GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    CAS 697-82-5
    Synonym Isopseudocumenol
    Boiling Point 230° to 231°C
    Molecular Formula C9H12O
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ALKYLPHENOLS, SOLID, N.O.S.
    Melting Point 92° to 95°C
    Odor Phenol-like
    UN Number UN2430
    Health Hazard 1 Danger
    Quantity 100g
    TSCA TSCA
    MDL Number MFCD00002228
    Beilstein 2042210
    Chemical Name or Material 2,3,5-Trimethylphenol
    Assay ≥98%

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P305+P351+P338-P309-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit