Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,5-Trimethylthiophene 98.0+%, TCI America™
SDP

Catalog No. T13271G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13271G 1 g
T13275G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13271G Supplier TCI America Supplier No. T13271G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1795-05-7
Molecular Formula C7H10S
Molecular Weight (g/mol) 126.22
MDL Number MFCD00142784
InChI Key QKZJQIHBRCFDGQ-UHFFFAOYSA-N
PubChem CID 74531
IUPAC Name 2,3,5-trimethylthiophene
SMILES CC1=CC(C)=C(C)S1

Specifications

Color Yellow
Boiling Point 82°C
Quantity 1 g
UN Number 1993
Formula Weight 126.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,3,5-Trimethylthiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.