Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trichlorophenol 98.0+%, TCI America™
SDP

Catalog No. T111410G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T111410G 10 g
T11141G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T111410G Supplier TCI America Supplier No. T111410G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 933-75-5
Molecular Formula C6H3Cl3O
Molecular Weight (g/mol) 197.439
MDL Number MFCD00002165
InChI Key XGCHAIDDPMFRLJ-UHFFFAOYSA-N
Synonym phenol, 2,3,6-trichloro, unii-ae2cmr564u, ccris 1937, ae2cmr564u, dsstox_cid_6209, dsstox_rid_78060, dsstox_gsid_26209, pubchem12938, acmc-209rle, 2,3,6-trichloro-phenol
PubChem CID 13618
IUPAC Name 2,3,6-trichlorophenol
SMILES C1=CC(=C(C(=C1Cl)O)Cl)Cl

Specifications

Melting Point 55°C
Boiling Point 250°C
Quantity 10 g
UN Number 2020
Formula Weight 197.44
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3,6-Trichlorophenol
EINECSNumber (3)-0931
RTECSNumber SN1300000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.