Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3,6-Trimethylphenol, MP Biomedicals

Catalog No. ICN222803
Click to view available options
Quantity:
10 g

Chemical Identifiers

CAS 2416-94-6
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00002229
InChI Key QQOMQLYQAXGHSU-UHFFFAOYSA-N
Synonym 3-hydroxypseudocumene, phenol, 2,3,6-trimethyl, 1-hydroxy-2,3,6-trimethylbenzene, 2,3,6-trimethyl-phenol, unii-05wkl2l5lj, 2,3,6-trimethyl phenol, 05wkl2l5lj, dsstox_cid_2187, dsstox_rid_76518, dsstox_gsid_22187
PubChem CID 17016
IUPAC Name 2,3,6-trimethylphenol
SMILES CC1=CC=C(C)C(O)=C1C

Specifications

Quantity 10 g
Formula Weight 136.19
Chemical Name or Material 2, 3, 6-Trimethyl Phenol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.