Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-(3-Benzoylphenyl)propionitrile 98.0+%, TCI America™
SDP

Catalog No. B293925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B293925G 25 g
B29395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B293925G Supplier TCI America Supplier No. B293925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 42872-30-0
Molecular Formula C16H13NO
Molecular Weight (g/mol) 235.29
MDL Number MFCD00044435
InChI Key RGYOCHMZSLUCNP-UHFFFAOYNA-N
Synonym 3-(1-Cyanoethyl)benzophenone, 3-Benzoyl-alpha-methylphenylacetonitrile
PubChem CID 39325
IUPAC Name 2-(3-benzoylphenyl)propanenitrile
SMILES CC(C#N)C1=CC=CC(=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C
Color White-Yellow
Quantity 25 g
Formula Weight 235.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2-(3-Benzoylphenyl)propionitrile
RTECSNumber CY1693000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.