Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Bis(2,6-diisopropylphenylimino)butane 98.0+%, TCI America™
SDP

Catalog No. B56061G
Click to view available options
Quantity:
200 mg
1 g

Chemical Identifiers

CAS 74663-77-7
Molecular Formula C28H40N2
Molecular Weight (g/mol) 404.642
MDL Number MFCD01862435
InChI Key YUFQUBWPYIPRHZ-UHFFFAOYSA-N
Synonym N,N′-Bis(2,6-diisopropylphenyl)butane-2,3-diimine
PubChem CID 608871
IUPAC Name 2-N,3-N-bis[2,6-di(propan-2-yl)phenyl]butane-2,3-diimine
SMILES CC(C)C1=C(C(=CC=C1)C(C)C)N=C(C)C(=NC2=C(C=CC=C2C(C)C)C(C)C)C

Specifications

Melting Point 101°C
Quantity 1 g
Formula Weight 404.64
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Bis(2,6-diisopropylphenylimino)butane
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.