Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-2,3-Butanediol, 98%

Catalog No. AAH6636936
Click to view available options
Quantity:
100 g
500 g

CAS: 513-85-9 | C4H10O2 | 90.122 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-85-9
Molecular Formula C4H10O2
Molecular Weight (g/mol) 90.122
MDL Number MFCD00004523
InChI Key OWBTYPJTUOEWEK-UHFFFAOYSA-N
Synonym 2,3-butanediol, 2,3-butylene glycol, 2,3-dihydroxybutane, dimethylethylene glycol, dimethylene glycol, pseudobutylene glycol, sym-dimethylethylene glycol, ccris 5501, dl-2,3-butanediol, 2,3-butanediol, r*,r*-.+/-.
PubChem CID 262
ChEBI CHEBI:62064
IUPAC Name butane-2,3-diol
SMILES CC(C(C)O)O

Specifications

Melting Point 25°C
Color Colorless to Yellow
Density 1.002 g/mL
Boiling Point 183°C to 184°C
Flash Point 85°C (185°F)
Odor Odorless
Refractive Index 1.433
Quantity 500 g
Beilstein 969165
Sensitivity Hygroscopic
Formula Weight 90.12
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (±)-2,3-Butanediol
Show More Show Less
Hazard Category H227-H320
Hazard Statement GHS H Statement
H227
Combustible liquid.
Precautionary Statement P210-P233-P240-P241-P242-P243-P264b-P280-P305+P351+P338-P374-P501c
EINECSNumber 208-173-6
RTECSNumber EK0532000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.