Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ (±)-2,3-Butanediol, 98%

Click to view available options
Quantity:
100g
500g
513-85-9
C4H10O2
90.122
MFCD00004523
OWBTYPJTUOEWEK-UHFFFAOYSA-N
2,3-butanediol,2,3-butylene glycol,2,3-dihydroxybutane,dimethylethylene glycol,dimethylene glycol,pseudobutylene glycol,sym-dimethylethylene glycol,ccris 5501,dl-2,3-butanediol,2,3-butanediol, r*,r*-.+/-.
262
CHEBI:62064
butane-2,3-diol
CC(C(C)O)O
513-85-9
C4H10O2
90.122
MFCD00004523
OWBTYPJTUOEWEK-UHFFFAOYSA-N
2,3-butanediol,2,3-butylene glycol,2,3-dihydroxybutane,dimethylethylene glycol,dimethylene glycol,pseudobutylene glycol,sym-dimethylethylene glycol,ccris 5501,dl-2,3-butanediol,2,3-butanediol, r*,r*-.+/-.
262
CHEBI:62064
butane-2,3-diol
CC(C(C)O)O
Sensitivity Hygroscopic
Boiling Point 183°C to 184°C
Quantity 100g
Odor Odorless
Melting Point 25°C
Density 1.002g/mL
Refractive Index 1.4330
Color Colorless to Yellow
Beilstein 969165
Formula Weight 90.12
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material (±)-2,3-Butanediol
Show More Show Less

GHS H Statement
H227
Combustible liquid.

GHS P Statement
P210-P280-P370+P378a-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, powder or water spray.
Store in a well-ventilated place.
Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : EK0532000

TSCA : TSCA