Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ 2,3-DCPE hydrochloride

Catalog No. 213710R
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-371-0R 10 mg
21-375-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 21-371-0R Supplier Tocris Bioscience™ Supplier No. 2137/10
This item is not returnable. View return policy

Selectively induces cancer cell apoptosis

2,3-DCPE hydrochloride is a selectively induces apoptosis and downregulates Bcl-XL protein expression in various human cancer cells versus normal cells in vitro. IC50 values are 0.89 and 12.6 μM for LoVo human colon cancer cell line and normal human fibroblasts respectively. Induces p21 expression and S-phase arrest in cancer cells via ERK-mediated pathways.

Specifications

Purity >99%
Quantity 10 mg
Product Type 2,3-DCPE hydrochloride
Molecular Weight (g/mol) 300.61
Inhibitors 2,3-DCPE hydrochloride

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.