Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Diaminotoluene, 97%

Catalog No. p-7022744
Click to view available options
Quantity:
5 g
25 g

CAS: 2687-25-4 | C7H10N2 | 122.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-25-4
Molecular Formula C7H10N2
Molecular Weight (g/mol) 122.17
MDL Number MFCD00011589
InChI Key AXNUJYHFQHQZBE-UHFFFAOYSA-N
Synonym 2,3-diaminotoluene, toluene-2,3-diamine, 3-methyl-o-phenylenediamine, 1,2-benzenediamine, 3-methyl, 3-methyl-1,2-phenylenediamine, 2,3-tolylenediamine, 2,3-toluylenediamine, 1,2-diamino-3-methylbenzene, 3-methyl-1,2-benzenediamine, 1-methyl-2,3-phenylenediamine
PubChem CID 17593
IUPAC Name 3-methylbenzene-1,2-diamine
SMILES CC1=CC=CC(N)=C1N

Specifications

Melting Point 56°C to 60°C
Boiling Point 225°C
Quantity 5 g
Formula Weight 122.17
Percent Purity 98%
Chemical Name or Material 2,3-Diaminotoluene
Hazard Category H301-H312+H332-H317-H319-H341-H350-H361f-H373
Hazard Statement GHS H Statement
H301-H312-H315-H319-H335
Toxic if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P201-P202-P260-P264b-P270-P271-P272-P280-P281-P301+P310-P302+P352-P304+P340-P305+P351+P338-P308+P313-P312-P330-P333+P313-P363-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 220-248-5
RTECSNumber XS9550000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.