Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde, 98%

Catalog No. AAB2114006
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2114006 5 g
AAB2114003 1 g
AAB2114014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2114006 Supplier Thermo Scientific Chemicals Supplier No. B2114006
Add to Cart

CAS: 2973-75-3 | C8H6Br2O3 | 309.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2973-75-3
Molecular Formula C8H6Br2O3
Molecular Weight (g/mol) 309.94
MDL Number MFCD00016978
InChI Key WKLKGSHBXNPUDU-UHFFFAOYSA-N
Synonym 5,6-dibromovanillin, acmc-1chm2, 2,3-dibromo-5-methoxy-4-hydroxybenzaldehyde, 2,3-bis bromanyl-5-methoxy-4-oxidanyl-benzaldehyde
PubChem CID 520452
IUPAC Name 2,3-dibromo-4-hydroxy-5-methoxybenzaldehyde
SMILES COC1=CC(C=O)=C(Br)C(Br)=C1O

Specifications

Melting Point 213°C to 217°C
Quantity 5 g
Beilstein 1968576
Sensitivity Air Sensitive
Formula Weight 309.94
Percent Purity 98%
Chemical Name or Material 2,3-Dibromo-4-hydroxy-5-methoxybenzaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.