Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dibromopropionyl chloride, 97%

Catalog No. p-4323218
Click to view available options
Quantity:
5 g
25 g

CAS: 18791-02-1 | C3H3Br2ClO | 250.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18791-02-1
Molecular Formula C3H3Br2ClO
Molecular Weight (g/mol) 250.31
MDL Number MFCD00000712
InChI Key HWKWYDXHMQQDQJ-UHFFFAOYNA-N
Synonym 2,3-dibromopropionyl chloride, propanoyl chloride, 2,3-dibromo, alpha,beta-dibromopropionyl chloride, propanoyl chloride,2,3-dibromo, propionyl chloride, 2,3-dibromo, acmc-209eqo, propanoyl chloride, 2,3-dibromo-9ci, 2,3-dibromopropanyl chloride, 3-02-00-00572 beilstein handbook reference
PubChem CID 86790
IUPAC Name 2,3-dibromopropanoyl chloride
SMILES ClC(=O)C(Br)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 2.1400g/mL
Boiling Point 193°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula BrCH2CH(Br)COCl
Quantity 5 g
Specific Gravity 2.14
Formula Weight 250.32
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dibromopropionyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Harmful if swallowed.
May be corrosive to metals.
Reacts violently with water.
Contact with water liberates toxic gas.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store in a dry place. Store in a closed container.
Keep only in original container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 242-575-2

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.