Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-1-propanol, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
616-23-9
C3H6Cl2O
128.99
MFCD00040446
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
2,3-dichloro-1-propanol,1-propanol, 2,3-dichloro,2,3-dichloropropanol,alpha,beta-dichlorohydrin,1,2-dichloro-3-propanol,glycerol alpha,beta-dichlorohydrin,2,3-dichloropropyl alcohol,glycerol 1,2-dichlorohydrin,2,3-dichloropropane-1-ol,ccris 954
12018
2,3-dichloropropan-1-ol
C(C(CCl)Cl)O
616-23-9
C3H6Cl2O
128.99
MFCD00040446
ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
2,3-dichloro-1-propanol,1-propanol, 2,3-dichloro,2,3-dichloropropanol,alpha,beta-dichlorohydrin,1,2-dichloro-3-propanol,glycerol alpha,beta-dichlorohydrin,2,3-dichloropropyl alcohol,glycerol 1,2-dichlorohydrin,2,3-dichloropropane-1-ol,ccris 954
12018
2,3-dichloropropan-1-ol
C(C(CCl)Cl)O
Boiling Point 182°C
Flash Point 93°C
Specific Gravity 1.35
Refractive Index 1.4835 to 1.4855
Quantity 5g
Infrared Spectrum Authentic
Density 1.3500g/mL
Color Colorless
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,acetone,ethanol
Formula Weight 128.99
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2, 3-Dichloro-1-propanol
Show More Show Less

GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Suspected of causing genetic defects.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:

GHS Signal Word: Danger

EINECSNumber : 210-470-