Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-1-propanol, 97%

Catalog No. p-3523289
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle

CAS: 616-23-9 | C3H6Cl2O | 128.99 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 616-23-9
Molecular Formula C3H6Cl2O
Molecular Weight (g/mol) 128.99
MDL Number MFCD00040446
InChI Key ZXCYIJGIGSDJQQ-UHFFFAOYSA-N
Synonym 2,3-dichloro-1-propanol, 1-propanol, 2,3-dichloro, 2,3-dichloropropanol, alpha,beta-dichlorohydrin, 1,2-dichloro-3-propanol, glycerol alpha,beta-dichlorohydrin, 2,3-dichloropropyl alcohol, glycerol 1,2-dichlorohydrin, 2,3-dichloropropane-1-ol, ccris 954
PubChem CID 12018
IUPAC Name 2,3-dichloropropan-1-ol
SMILES C(C(CCl)Cl)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.3500g/mL
Boiling Point 182°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.4835 to 1.4855
Quantity 5 g
Specific Gravity 1.35
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in ether,acetone,ethanol
Formula Weight 128.99
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 3-Dichloro-1-propanol
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Fatal in contact with skin.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN:
EINECSNumber 210-470-

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.