Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-5-fluoropyridine, 98%, Thermo Scientific™

Catalog No. AAH6437303
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6437303 1 g
AAH64373MD 250 mg
AAH6437306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH6437303 Supplier Thermo Scientific Chemicals Supplier No. H6437303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2,3-Dichloro-5-fluoropyridine is a reagent that is used in the synthesis of 3-Chloro-5-fluoro Clodinafop Propargyl (C368645), which is an impurity of Clodinafop Propargyl.

Solubility
Slightly soluble in chloroform, methanol and water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 185985-40-4
Molecular Formula C5H2Cl2FN
Molecular Weight (g/mol) 165.976
MDL Number MFCD08669666
InChI Key YIYIQENCPIISLL-UHFFFAOYSA-N
PubChem CID 18520008
IUPAC Name 2,3-dichloro-5-fluoropyridine
SMILES C1=C(C=NC(=C1Cl)Cl)F

Specifications

Quantity 1 g
Beilstein 13831996
Synonym pyridine,2,3-dichloro-5-fluoro, pyridine, 2,3-dichloro-5-fluoro, abbypharma ap-31-5779, 2,3-dichloro-5-fluoro-pyridine
Solubility Information Slightly soluble in chloroform,methanol and water.
Formula Weight 165.98
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2,3-Dichloro-5-fluoropyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.