Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloro-6-nitroquinoxaline 98.0+%, TCI America™
SDP

Catalog No. D47571G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D47571G 1 g
D47575G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D47571G Supplier TCI America Supplier No. D47571G

Chemical Identifiers

CAS 2379-60-4
Molecular Formula C8H3Cl2N3O2
Molecular Weight (g/mol) 244.03
MDL Number MFCD00454905
InChI Key SFJCUOAQTGDBPO-UHFFFAOYSA-N
PubChem CID 689090
IUPAC Name 2,3-dichloro-6-nitroquinoxaline
SMILES [O-][N+](=O)C1=CC=C2N=C(Cl)C(Cl)=NC2=C1

Specifications

Melting Point 153°C
Color White-Yellow
Quantity 1 g
Formula Weight 244.03
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloro-6-nitroquinoxaline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.