Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichlorobenzaldehyde, 99%

Catalog No. p-3778189
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle

CAS: 6334-18-5 | C7H4Cl2O | 175.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6334-18-5
Molecular Formula C7H4Cl2O
Molecular Weight (g/mol) 175.01
MDL Number MFCD00010127
InChI Key LLMLNAVBOAMOEE-UHFFFAOYSA-N
Synonym benzaldehyde, 2,3-dichloro, dichlorobenzaldehyde, benzaldehyde, dichloro, ccris 8612, dichlorbenzaldehyd, pubchem3079, 2,3-dichlorobenzaldehyd, benzaldehyde,3-dichloro, 2,3-dichloro-benzaldehyde, acmc-209nf0
PubChem CID 35745
IUPAC Name 2,3-dichlorobenzaldehyde
SMILES ClC1=CC=CC(C=O)=C1Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 60.0°C to 64.0°C
Color White
Density 135g/mL
Boiling Point 144.0°C (30.0 mmHg)
Flash Point 135°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Cl2C6H3CHO
Quantity 25 g
Beilstein 07, III, 878
Solubility Information Solubility in water: slightly soluble
Formula Weight 175.01
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2, 3-Dichlorobenzaldehyde
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 228-711-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.