Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloropropionic Acid 98.0+%, TCI America™
SDP

Catalog No. D040725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D040725G 25 g
D04075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D040725G Supplier TCI America Supplier No. D040725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 565-64-0
Molecular Formula C3H4Cl2O2
Molecular Weight (g/mol) 142.963
MDL Number MFCD00021695
InChI Key GKFWNPPZHDYVLI-UHFFFAOYSA-N
PubChem CID 11263
IUPAC Name 2,3-dichloropropanoic acid
SMILES C(C(C(=O)O)Cl)Cl

Specifications

Melting Point 52°C
Color White-Yellow
Boiling Point 210°C
Quantity 25 g
UN Number 2923
Formula Weight 142.96
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dichloropropionic Acid
RTECSNumber UF0700000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.