Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloropyrazine, 98%

Catalog No. AC377040250
Click to view available options
Quantity:
5 g
25 g
100 g

CAS: 4858-85-9 | C4H2Cl2N2 | 148.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4858-85-9
Molecular Formula C4H2Cl2N2
Molecular Weight (g/mol) 148.97
MDL Number MFCD00040964
InChI Key MLCNOCRGSBCAGH-UHFFFAOYSA-N
Synonym pyrazine, 2,3-dichloro, 2,3-dichloro-pyrazine, 2,3-dichloro pyrazine, 2,3-dichloro-1,4-diazine, 2,3-dichlorpyrazin, dichloropyrazine, dichloro pyrazine, zlchem 169, 2 3-dichloropyrazine, 2,3 dichloropyrazine
PubChem CID 78575
IUPAC Name 2,3-dichloropyrazine
SMILES ClC1=NC=CN=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 22.5°C to 24.5°C
Density 1.4310g/mL
Boiling Point 105.0°C to 107.0°C (50.0 mmHg)
Flash Point 102°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Gravity 1.431
Solubility Information Solubility in water: insoluble
Formula Weight 148.98
Percent Purity 98%
Chemical Name or Material 2, 3-Dichloropyrazine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a
EINECSNumber 225-460-1

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.