Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloropyrazine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
4858-85-9
C4H2Cl2N2
148.98
MFCD00040964
MLCNOCRGSBCAGH-UHFFFAOYSA-N
pyrazine, 2,3-dichloro,2,3-dichloro-pyrazine,2,3-dichloro pyrazine,2,3-dichloro-1,4-diazine,2,3-dichlorpyrazin,dichloropyrazine,dichloro pyrazine,zlchem 169,2 3-dichloropyrazine,2,3 dichloropyrazine
78575
2,3-dichloropyrazine
C1=CN=C(C(=N1)Cl)Cl
4858-85-9
C4H2Cl2N2
148.98
MFCD00040964
MLCNOCRGSBCAGH-UHFFFAOYSA-N
pyrazine, 2,3-dichloro,2,3-dichloro-pyrazine,2,3-dichloro pyrazine,2,3-dichloro-1,4-diazine,2,3-dichlorpyrazin,dichloropyrazine,dichloro pyrazine,zlchem 169,2 3-dichloropyrazine,2,3 dichloropyrazine
78575
2,3-dichloropyrazine
C1=CN=C(C(=N1)Cl)Cl
Specific Gravity 1.431
Flash Point 102°C
Density 1.4310g/mL
Melting Point 22.5°C to 24.5°C
Boiling Point 105.0°C to 107.0°C (50.0 mmHg)
Quantity 100g
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Solubility Information Solubility in water: insoluble
Formula Weight 148.98
Percent Purity 98%
Chemical Name or Material 2, 3-Dichloropyrazine
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 225-460-1