Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dichloroquinoxaline, 97%

Catalog No. ac330830100
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle

Catalog No. AC330830100

Supplier: Thermo Scientific Chemicals 330830100

Only null left
Add to Cart

CAS: 2213-63-0 | C8H4Cl2N2 | 199.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2213-63-0
Molecular Formula C8H4Cl2N2
Molecular Weight (g/mol) 199.04
MDL Number MFCD00006720
InChI Key SPSSDDOTEZKOOV-UHFFFAOYSA-N
Synonym quinoxaline, 2,3-dichloro, 2,3-dichloro-quinoxaline, ccris 4789, quinoxaline, dichloro, dichloroquinoxaline, pubchem15473, 2,3 dichloroquinoxaline, 2.3-dichloroquinoxaline, acmc-1cfck, 3,4-dichloroquinoxaline
PubChem CID 16659
IUPAC Name 2,3-dichloroquinoxaline
SMILES C1=CC=C2C(=C1)N=C(C(=N2)Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 150°C to 155°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Quantity 10 g
Beilstein 23, 177
Formula Weight 199.04
Percent Purity 97%
Physical Form Needles
Chemical Name or Material 2, 3-Dichloroquinoxaline, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
Causes skin irritation.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 218-667-3
RTECSNumber VD1720000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.