Learn More
Thermo Scientific Chemicals 2',3'-Dideoxycytidine, 98+%
Description
2',3'-Dideoxycytidine in cells, it is phosphorylated to the active triphosphate form, ddCTP, which acts as a substrate for HIV reverse transcriptase (Ki = 51 nM). It incorporates into viral DNA where it terminates chain elongation when the missing hydroxyl group is encountered. It is also is used as a DNA chain-terminating nucleotide for DNA sequencing methods based on the Sanger chain-termination method.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 216°C to 220°C |
| Packaging | Glass bottle |
| Quantity | 250 mg |
| Beilstein | 654956 |
| Solubility Information | Soluble in water (76.4mg/ml at 25°C),DMSO (42mg/ml at 25°C),and ethanol (<1mg/ml at 25°C). |
| IUPAC Name | 4-amino-1-[(2R,5S)-5-(hydroxymethyl)oxolan-2-yl]pyrimidin-2-one |
| Formula Weight | 211.22 |
| Percent Purity | ≥98% |
| Physical Form | Solid |
| Chemical Name or Material | 2′,3′-Dideoxycytidine |
Safety and Handling
| Hazard Category | H351 |
| Precautionary Statement | P201-P202-P281-P308+P313-P501c |
| RTECSNumber | HA3870000 |
| TSCA | No |
| Recommended Storage | Keep cold |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.