Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Difluorophenylboronic Acid (contains varying amounts of Anhydride), TCI America™
SDP

Catalog No. p-7193929 Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
5 g
25 g

Specifications

Chemical Name or Material 2,3-Difluorophenylboronic Acid (contains varying amounts of Anhydride)
CAS 121219-16-7
Color White-Yellow
Molecular Formula C6H5BF2O2
Quantity 5 g
MDL Number MFCD01863170
Synonym 2,3-difluorobenzeneboronic acid, 2,3-difluorophenyl boronic acid, 2,3-difluorophenyl boranediol, 2,3-difluorophenylboronicacid, 2,3-difluoro phenylboric acid, 2,3-fluorobenzene boronic acid, 2,3-difluoro phenylboronic acid, 2,3-difluorophenyloboric acid, boronic acid, 2,3-difluorophenyl
InChI Key SZYXKFKWFYUOGZ-UHFFFAOYSA-N
SMILES B(C1=C(C(=CC=C1)F)F)(O)O
IUPAC Name (2,3-difluorophenyl)boronic acid
Molecular Weight (g/mol) 157.911
PubChem CID 2734333
Formula Weight 157.91
Physical Form Crystalline Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.