Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole 98.0+%, TCI America™
SDP

Catalog No. D51625G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
1 g
5 g

Chemical Identifiers

CAS 344928-74-1
Molecular Formula C16H17N
Molecular Weight (g/mol) 223.319
InChI Key XQWHKLIJFAHNNR-UHFFFAOYSA-N
PubChem CID 10751584
IUPAC Name 1,1,3-trimethyl-2-methylidenebenzo[e]indole
SMILES CC1(C(=C)N(C2=C1C3=CC=CC=C3C=C2)C)C

Specifications

Melting Point 116°C
Quantity 5 g
Formula Weight 223.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydro-1,1,3-trimethyl-2-methylene-1H-benzo[e]indole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.