Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2,3-Dihydro-7-azaindole, ≥97%

Click to view available options
Quantity:
5g
25g
100g
10592-27-5
C7H8N2
120.155
MFCD06659751
ZFFYPGZDXUPKNK-UHFFFAOYSA-N
2,3-dihydro-1h-pyrrolo 2,3-b pyridine,7-azaindoline,2,3-dihydro-7-azaindole,1h,2h,3h-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 2,3-dihydro,2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline,2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline,pubchem18022,acmc-2098hy,ksc492q5f
10011889
2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
C1CNC2=C1C=CC=N2
10592-27-5
C7H8N2
120.155
MFCD06659751
ZFFYPGZDXUPKNK-UHFFFAOYSA-N
2,3-dihydro-1h-pyrrolo 2,3-b pyridine,7-azaindoline,2,3-dihydro-7-azaindole,1h,2h,3h-pyrrolo 2,3-b pyridine,1h-pyrrolo 2,3-b pyridine, 2,3-dihydro,2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline,2,3-dihydro-1h-pyrrolo 2,3-b pyridine 7-azaindoline,pubchem18022,acmc-2098hy,ksc492q5f
10011889
2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
C1CNC2=C1C=CC=N2
Applications
It can be used as pharmaceutical raw material and intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Quantity 100g
Color White
Melting Point 78°C to 83°C
Beilstein 116366
Formula Weight 120.15
Physical Form Powder
Percent Purity ≥97%
Chemical Name or Material 2,3-Dihydro-7-azaindole

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning