Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride, 97%

Catalog No. AAH3357906
Click to view available options
Quantity:
250 mg
5 g

CAS: 115010-11-2 | C8H7ClO3S | 218.65 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115010-11-2
Molecular Formula C8H7ClO3S
Molecular Weight (g/mol) 218.65
MDL Number MFCD02677703
InChI Key RVWYPBARHGPULM-UHFFFAOYSA-N
Synonym 2,3-dihydrobenzofuran-5-sulfonyl chloride, 2,3-dihydro-1-benzofuran-5-sulfonoylchloride, 2,3-dihydro-1-benzofuran-5-sulfonoyl chloride, 2,3-dihydrobenzofuran-5-sulfonylchloride, 2,3-dihydro-benzofuran-5-sulfonyl chloride, 2,3-dihydrobenzo b furan-5-sulfonyl chloride, pubchem14874, 5-benzofuransulfonylchloride, 2,3-dihydro, 2,3-dihydro-1-benzofuran-5-sulfonylchloride, chloro-2,3-dihydrobenzo b furan-5-ylsulfone
PubChem CID 2776154
IUPAC Name 2,3-dihydro-1-benzofuran-5-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC2=C(OCC2)C=C1

Specifications

Melting Point 83°C to 85°C
Quantity 5 g
UN Number UN3261
Formula Weight 218.66
Percent Purity 97%
Chemical Name or Material 2,3-Dihydrobenzo[b]furan-5-sulfonyl chloride
Hazard Category H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.