Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydroxybenzaldehyde 98.0+%, TCI America™
SDP

Catalog No. D147825G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D147825G 25 g
D14785G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D147825G Supplier TCI America Supplier No. D147825G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 24677-78-9
Molecular Formula C7H6O3
Molecular Weight (g/mol) 138.122
MDL Number MFCD00003324
InChI Key IXWOUPGDGMCKGT-UHFFFAOYSA-N
Synonym o-pyrocatechualdehyde, 5,6-dihydroxybenzaldehyde, benzaldehyde, 2,3-dihydroxy, benzaldehyde, dihydroxy, 2,3-dihydroxybenzaldehyde, dihydroxybenzaldehyde, pubchem6561, acmc-1clux, 2,3-dihydroxy benzaldehyde, 2,3-dihydroxy-benzaldehyde
PubChem CID 90579
ChEBI CHEBI:50197
IUPAC Name 2,3-dihydroxybenzaldehyde
SMILES C1=CC(=C(C(=C1)O)O)C=O

Specifications

Melting Point 107°C
Boiling Point 120°C
Quantity 25 g
Formula Weight 138.12
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxybenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.