Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dihydroxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C005425G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 303-38-8
Molecular Formula C7H6O4
Molecular Weight (g/mol) 154.121
MDL Number MFCD00002446
InChI Key GLDQAMYCGOIJDV-UHFFFAOYSA-N
Synonym pyrocatechuic acid, o-pyrocatechuic acid, 2-pyrocatechuic acid, 3-hydroxysalicylic acid, dobk, dhba, 2,3-dihydroxybenzoicacid, benzoic acid, 2,3-dihydroxy, catecholcarboxylic acid, 2,3-dihydroxy-benzoic acid
PubChem CID 19
ChEBI CHEBI:18026
IUPAC Name 2,3-dihydroxybenzoic acid
SMILES C1=CC(=C(C(=C1)O)O)C(=O)O

Specifications

Melting Point 205°C
Quantity 25 g
Formula Weight 154.12
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,3-Dihydroxybenzoic Acid
EINECSNumber (3)-1616
RTECSNumber DG8576490
TSCA No
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.