Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimercapto-1-propanol, 97.0-100.5%, ACROS Organics™

Click to view available options
Quantity:
5mL
25mL
Packaging:
Glass bottle
59-52-9
C3H8OS2
124.216
MFCD00004864
WQABCVAJNWAXTE-UHFFFAOYSA-N
dimercaprol,2,3-dimercapto-1-propanol,2,3-dimercaptopropanol,dithioglycerine,dicaptol,sulfactin,dimercaptopropanol,british anti-lewisite,2,3-dithiopropanol,antoxol
3080
CHEBI:64198
2,3-bis(sulfanyl)propan-1-ol
C(C(CS)S)O
59-52-9
C3H8OS2
124.216
MFCD00004864
WQABCVAJNWAXTE-UHFFFAOYSA-N
dimercaprol,2,3-dimercapto-1-propanol,2,3-dimercaptopropanol,dithioglycerine,dicaptol,sulfactin,dimercaptopropanol,british anti-lewisite,2,3-dithiopropanol,antoxol
3080
CHEBI:64198
2,3-bis(sulfanyl)propan-1-ol
C(C(CS)S)O
Flash Point >112°C
Infrared Spectrum Authentic
Color Colorless to Yellow
Specific Gravity 1.243
Refractive Index 1.5700 to 1.5740
Boiling Point 120.0°C (15.0 mmHg)
Packaging Glass bottle
Quantity 5mL
Density 1.2430g/mL
CAS Min % 97.0
CAS Max % 100.0
Assay Percent Range 97.0-100.5%
Linear Formula HSCH2CH(SH)CH2OH
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol and vegetable oils
Formula Weight 124.22
Physical Form Liquid
Percent Purity 97.0 to 100.5%
Chemical Name or Material 2,3-Dimercapto-1-propanol, 97.0-100.5%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Wear eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Warning

EINECSNumber : 200-433-7

RTECSNumber : UB2625000

TSCA : TSCA