Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,3-Dimercapto-1-propanol, 97.0-100.5%

Catalog No. AC115300250
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle

CAS: 59-52-9 | C3H8OS2 | 124.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59-52-9
Molecular Formula C3H8OS2
Molecular Weight (g/mol) 124.22
MDL Number MFCD00004864
InChI Key WQABCVAJNWAXTE-UHFFFAOYSA-N
Synonym dimercaprol, 2,3-dimercapto-1-propanol, 2,3-dimercaptopropanol, dithioglycerine, dicaptol, sulfactin, dimercaptopropanol, british anti-lewisite, 2,3-dithiopropanol, antoxol
PubChem CID 3080
ChEBI CHEBI:64198
IUPAC Name 2,3-bis(sulfanyl)propan-1-ol
SMILES C(C(CS)S)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2430g/mL
Boiling Point 120.0°C (15.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 97.0-100.5%
Packaging Glass bottle
Refractive Index 1.5700 to 1.5740
Linear Formula HSCH2CH(SH)CH2OH
Quantity 25 mL
Merck Index 15,3231
Specific Gravity 1.243
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in alcohol and vegetable oils
Formula Weight 124.22
Percent Purity 97.0-100.5%
Physical Form Liquid
Chemical Name or Material 2,3-Dimercapto-1-propanol, 97.0-100.5%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation or rash occurs: Get medical advice/attention.
If eye irritation persists: Get med
EINECSNumber 200-433-7
RTECSNumber UB2625000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.